Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ahsa1, Rat, Clone: 25F2.D10, Abnova™

Catalog No. 89112995 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-995 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-995 Supplier Abnova Corporation Supplier No. MAB6669
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Ahsa1.

Specifications

Antigen Ahsa1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 25F2.D10
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Ahsa1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2
Gene Ahsa1
Gene Alias BC023857/MGC36589/MGC36618/p38
Gene Symbols Ahsa1
Host Species Rat
Immunogen Mouse Aha1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217737
Test Specificity Approximately 38 KDa, can run at up to 45 KDa on SDS-PAGE.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.