Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AID Rat anti-Human, Alexa Fluor 647, Clone: EK2-5G9, BD

Catalog No. BDB565785 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565785 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565785 Supplier BD Biosciences Supplier No. 565785
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The EK2-5G9 monoclonal antibody specifically binds to human Activation-induced cytidine deaminase (AICDA), also known as AID, or Single-stranded DNA cytosine deaminase. AID is an enzyme that preferentially acts upon certain DNA sequence motifs, such as those located in immunoglobulin variable regions, to induce cytidine-to-uracil mutations. AID-induced mutations initiate both somatic hypermutation (SHM) and class switch recombination (CSR) of the immunoglobulin genes in activated B cells during immune responses. While essential for immunoglobulin gene diversification, dysregulated AID activity can result in genomic instability and oncogenic transformation. Originally thought to be B-cell specific, AID now appears to be abnormally expressed in several epithelial cancers. AID may also play roles in DNA demethylation.

TRUSTED_SUSTAINABILITY

Specifications

Antigen AID
Applications Flow Cytometry, Immunofluorescence, Immunocytochemistry
Classification Monoclonal
Clone EK2-5G9
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Description AID; ARP2; CDA2; HEL-S-284; HIGM2; Cytidine aminohydrolase
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Human AID (185-198) Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.