Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apc, Rat, Clone: KT45, Abnova™

Catalog No. 89110577 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-577 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-577 Supplier Abnova Corporation Supplier No. MAB1683
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Apc.

Specifications

Antigen Apc
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone KT45
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Apc.
Dilution Western Blot (1:500-1:5000) Immunofluorescence (1:100-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Apc
Gene Alias AI047805/AU020952/AW124434/Min
Gene Symbols Apc
Host Species Rat
Immunogen Recombinant protein corresponding to amino acids 2170-2394 of mouse Apc.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11789
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.