Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl2l11, Rat, Clone: 14A8, Abnova™

Catalog No. 89111546 Shop All Abnova Corporation Products
missing translation for 'changeView'
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-546 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. 89-111-546 Supplier Abnova Corporation missing translation for 'supplierNo' MAB3704
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Bcl2l11.

Specifications

Antigen Bcl2l11
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 14A8
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Bcl2l11.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Bcl2l11
Gene Alias 1500006F24Rik/Bim/Bod
Gene Symbols Bcl2l11
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Bcl2l11.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12125
Test Specificity Recognizes human, mouse and monkey BimL, BimEL and BimS.
Target Species Human, Monkey, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.