Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid-stimulating hormone (TSH), Rat, Clone: 12E10, Abnova™

Catalog No. 89111545 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-545 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-545 Supplier Abnova Corporation Supplier No. MAB3703
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Bmf.

Specifications

Antigen Thyroid-stimulating hormone (TSH)
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 12E10
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Bmf.
Dilution Western Blot (0.5-1.0 ug/mL) Flow Cytometry (1:100- 1:1000) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Bmf
Gene Alias AW260063/AW411872
Gene Symbols Bmf
Host Species Rat
Immunogen Recombinant GST fusion protein corresponding to mouse Bmf.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 171543
Test Specificity Recognizes human and mouse Bmf.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.