Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casp1, Rat, Clone: 1H11, Abnova™

Catalog No. 89111411 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-411 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-411 Supplier Abnova Corporation Supplier No. MAB3427
Add to Cart
Add to Cart

Rat monoclonal antibody raised against synthetic peptide of Casp1.

Sequence: TALEMVKEVKEFAAC

Specifications

Antigen Casp1
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1H11
Conjugate Unconjugated
Description Rat monoclonal antibody raised against synthetic peptide of Casp1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Casp1
Gene Alias ICE/Il1bc
Gene Symbols Casp1
Host Species Rat
Immunogen A synthetic peptide corresponding to amino acids 206-220 of mouse Casp1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12362
Test Specificity Recognizes an epitope within the p20 fragment of mouse caspase-1. Detects a band of ∼45 KDa by Western blot. Does not cross-react with mouse caspase-11 or -12.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.