Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ccr9, Rat, Clone: 7E7, Abnova™

Catalog No. 89111005 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-005 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-005 Supplier Abnova Corporation Supplier No. MAB2435
Add to Cart
Add to Cart

Rat monoclonal antibody raised against synthetic peptide of Ccr9.

Specifications

Antigen Ccr9
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 7E7
Conjugate Unconjugated
Description Rat monoclonal antibody raised against synthetic peptide of Ccr9.
Dilution The optimal working dilution should be determined by the end user.
Formulation In tissue culture supernatant (0.09% sodium azide)
Gene Ccr9
Gene Alias Cmkbr10/GPR-9-6
Gene Symbols Ccr9
Host Species Rat
Immunogen A synthetic peptide corresponding to amino acids 322 of mouse Ccr9.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12769
Test Specificity Murine Ccr9.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.