Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD105 Rat anti-Mouse, Brilliant Violet 421, Clone: MJ7/18, BD

Catalog No. BDB562760 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562760 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562760 Supplier BD Biosciences Supplier No. 562760
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The MJ7/18 monoclonal antibody specifically binds to mouse CD105 (also known as endoglin) which is a homodimer of 90-kDa subunits and is predominantly expressed on vascular endothelial cells. High levels of mouse endoglin mRNA have been reported to be detectable in the ovary, uterus, NCTC-2071 fibroblasts, and to a lesser extent, in heart, muscle and stromal cells in connective tissue of various organs. Endoglin has been reported to play an essential role in embryonic angiogenesis. Both mouse and human endoglin display strong amino-acid sequence homology to the transmembrane and cytoplasmic regions of the type III TGF-ß receptor.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD105
Applications Flow Cytometry
Classification Monoclonal
Clone MJ7/18
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Endoglin; Edg; EGLN; Eng; MJ7/18 antigen; S-endoglin
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse skin (inflamed)
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.