Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-CD122, Clone: 5H4,

Catalog No. MABF840MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF840MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF840MI Supplier MilliporeSigma™ Supplier No. MABF840
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects CD122 clone: 5H4 in Mouse samples, and it is validated for Flow Cytometry

The interleukin-2 receptor (IL-2R) is a heterotrimeric membrane receptor composed of alpha, beta, and gamma subuits. The alpha and beta chains are involved in IL-2 binding, while the beta and gamma chain mediate cellular signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD122
Applications Flow Cytometry
Classification Monoclonal
Clone 5H4
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P16297
Gene Symbols Il2rb
Host Species Rat
Immunogen Whole cells corresponding to mouse CD122/IL-2R beta.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_032394
Target Species Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.