Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD127 Rat anti-Mouse, PE, Clone: SB/199, BD

Catalog No. BDB552543 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552543 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552543 Supplier BD Biosciences Supplier No. 552543
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The SB/199 monoclonal antibody specifically binds to mouse CD127, the 65-75 kDa type-I transmembrane protein IL-7Rα. The high affinity IL-7 receptor complex is composed of at least two transmembrane proteins, IL-7Rα and CD132, the common γ chain. CD127 has some sequence homology to the cytokine receptor superfamily (also known as the hematopoietin receptor superfamily). Micelacking CD127 display profoundly impaired development of the B and T lymphoid cell lineages, but display no obvious non-lymphoid abnormalities. IL-7Rα is expressed on common lymphoid progenitors and early stages of B lineage development in the bone marrow, on the earliest thymocyte progenitors, on CD4-CD8- double-negative and CD4+ and CD8+ single-positive thymocytes, and on most peripheral T lymphocytes. Intestinal intraepithelial lymphocytes with low-density γδ TCR upregulate CD127 expression in response to IL-2, which may be secreted by neighboring αβ TCR-bearing T cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD127
Applications Flow Cytometry
Classification Monoclonal
Clone SB/199
Concentration 0.2mg/mL
Conjugate PE
Description Interleukin-7 receptor alpha chain; IL-7R alpha; IL-7RA; IL-7Rα; Il7r
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen BALB/c mouse pre-B cell line 1A9
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.