Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd274, Rat, Clone: MIH5, Abnova™

Catalog No. 89109218 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-218 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-218 Supplier Abnova Corporation Supplier No. MAB11003

Rat monoclonal antibody raised against Cd274.

Specifications

Antigen Cd274
Applications Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone MIH5
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Cd274.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Cd274
Gene Alias B7-H1/PD-L1/Pdcd1l1/Pdcd1lg1
Gene Symbols Cd274
Host Species Rat
Immunogen Mouse Cd274.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60533
Test Specificity The monoclonal antibody reacts with mouse Cd274, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.