Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD281 (TLR1) Rat anti-Mouse, Brilliant Violet 421, Clone: TR23, BD

Catalog No. BDB565792 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565792 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565792 Supplier BD Biosciences Supplier No. 565792
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The TR23 monoclonal antibody specifically binds to CD281, which is also known as Toll-like receptor 1 (TLR1), or Toll/interleukin-1 receptor-like protein (TIL). CD281 is an ∽98 kDa type I transmembrane glycoprotein that belongs to the Toll-like receptor (TLR) family. CD281 functions as a pattern recognition receptor (PRR) for pathogen-associated molecular patterns (PAMP) that are broadly expressed by microbial pathogens and plays essential roles in innate and adaptive immunity. In the process of binding microbial ligands, such as triacylated microbial lipopeptides, TLR1 associates with CD282 (TLR2). CD281 is expressed on B cells, monocytes/macrophages, and dendritic cells. CD281/TLR1-deficient mice display impaired production of inflammatory cytokines in response to microbial ligands.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD281 (TLR1)
Applications Flow Cytometry
Classification Monoclonal
Clone TR23
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Cd281; Tlr1; Toll-like receptor 1; TIL; Toll/interleukin-1 receptor-like
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse TLR1 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.