Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd3e, Rat, Clone: CD3-H5, Abnova™

Catalog No. 89110373 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-373 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-373 Supplier Abnova Corporation Supplier No. MAB1353
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Cd3e.

Specifications

Antigen Cd3e
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone CD3-H5
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Cd3e.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Cd3e
Gene Alias AI504783/CD3/CD3epsilon/T3e
Gene Symbols Cd3e
Host Species Rat
Immunogen Native from Con A-activated mouse LN cells.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12501
Test Specificity React with mouse CD3.
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.