Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd4, Rat, Clone: JXYT4, Abnova™

Catalog No. 89112826 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-826 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-826 Supplier Abnova Corporation Supplier No. MAB6354
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Cd4.

Specifications

Antigen Cd4
Applications ELISA
Classification Monoclonal
Clone JXYT4
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Cd4.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene Cd4
Gene Alias L3T4/Ly-4
Gene Symbols Cd4
Host Species Rat
Immunogen Mouse T-lymphocytes.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12504
Test Specificity This antibody react with murine T-cell surface antigen Cd4 marker that is expressed by the helper/inducer subset of mouse T cells.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.