Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd4, Rat, PE, Clone: GK1.5, Abnova™

Catalog No. 89112136 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-136 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-136 Supplier Abnova Corporation Supplier No. MAB5127
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Cd4.

Specifications

Antigen Cd4
Applications Flow Cytometry
Classification Monoclonal
Clone GK1.5
Concentration 0.5 mg/mL
Conjugate PE
Description Rat monoclonal antibody raised against native Cd4.
Dilution Flow Cytometry (3 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene Cd4
Gene Alias L3T4/Ly-4
Gene Symbols Cd4
Host Species Rat
Immunogen Mouse CTL clone V4 cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12504
Test Specificity This antibody reacts with an extracellular epitope of mouse CD4 transmembrane glycoprotein (55 KDa).
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.