Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD66aCD, Rat, Clone: YTH71.3, Abnova™

Catalog No. 89135926 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-926 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-926 Supplier Abnova Corporation Supplier No. MAB5263
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native CD66aCD.

Specifications

Antigen CD66aCD
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone YTH71.3
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native CD66aCD.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Rat
Immunogen Native purified CD66aCD from human lymphocytes.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity YTH71.3 recognizes human CD66acd. CD66acd is expressed by human neutrophils and cells in some cases of colon carcinoma. Clone YTH71.3 reacts with transfectants containing BGPa, NCA and CGM1a which are CEA subfamily members(2).
Target Species Human, Monkey
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.