Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd70, Rat, Clone: FR70, Abnova™

Catalog No. 89109160 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-160 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-160 Supplier Abnova Corporation Supplier No. MAB10947

Rat monoclonal antibody raised against Cd70.

Specifications

Antigen Cd70
Applications Flow Cytometry, Functional Assay, Immunoprecipitation
Classification Monoclonal
Clone FR70
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Cd70.
Dilution The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide)
Gene Cd70
Gene Alias CD27LG/Cd27l/Tnfsf7
Gene Symbols Cd70
Host Species Rat
Immunogen Mouse Cd70.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21948
Test Specificity The monoclonal antibody reacts with mouse CD70, a 30-33 kDa type II transmembrane glycoprotein.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.