Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd80, Rat, Clone: MsxhB7-1, Abnova™

Catalog No. 89110363 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-363 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-363 Supplier Abnova Corporation Supplier No. MAB1326
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Cd80.

Specifications

Antigen Cd80
Applications ELISA
Classification Monoclonal
Clone MsxhB7-1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Cd80.
Dilution ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Cd80
Gene Alias B7-1/B7.1/Cd28l/Ly-53/Ly53/MIC17/TS/A-1
Gene Symbols Cd80
Host Species Rat
Immunogen Native purified Cd80 from mouse LPS-activated B cells.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12519
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.