Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd86, Rat, FITC, Clone: GL-1, Abnova™

Catalog No. 89113808 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-808 Supplier Abnova Corporation Supplier No. MAB9824
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Cd86.

Specifications

Antigen Cd86
Applications Flow Cytometry
Classification Monoclonal
Clone GL-1
Concentration 0.5 mg/mL
Conjugate FITC
Description Rat monoclonal antibody raised against Cd86.
Dilution Flow Cytometry (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Cd86
Gene Alias B7/B7-2/B7.2/B70/CLS1/Cd28l2/ETC-1/Ly-58/Ly58/MB7/MB7-2/TS/A-2
Gene Symbols Cd86
Host Species Rat
Immunogen LPS-activated CBA/Cs mouse splenic B cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12524
Test Specificity The rat monoclonal antibody GL-1 reacts with Cd86 (B7-2), a 70-80 kDa type I transmembrane glycoprotein of immunoglobulin supergene family, expressed on professional antigen-presenting cells, such as dendritic cells, macrophages or activated B lymphocytes.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.