Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd8a, Rat, PE-TXRD, Clone: 53-6.7, Abnova™

Catalog No. 89112706 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-706 Supplier Abnova Corporation Supplier No. MAB6108
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Cd8a.

Specifications

Antigen Cd8a
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 53-6.7
Conjugate PE-Texas Red
Description Rat monoclonal antibody raised against native Cd8a.
Dilution Flow Cytometry (0.2 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Cd8a
Gene Alias BB154331/Ly-2/Ly-35/Ly-B/Lyt-2
Gene Symbols Cd8a
Host Species Rat
Immunogen Native purified from mouse thymus/spleen cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12525
Test Specificity Specificity alpha chain of the murine CD8 heterodimer, Mr 32-34 KDa
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.