Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD96 Rat anti-Mouse, Brilliant Violet 421, Clone: 6A6, BD

Catalog No. BDB565687 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565687 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565687 Supplier BD Biosciences Supplier No. 565687
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6A6 monoclonal antibody specifically binds to mouse CD96, which is also known as T cell-activated increased late expression protein (Tactile, TACT). CD96 is a type I transmembrane glycoprotein that belongs to the Ig gene superfamily. It is variably expressed on NK cells , and T cells at all developmental stages, including thymocytes and mature αβ T cells, γδ T cells , and NKT cells. CD96 interacts with Nectin-1 (CD111) and CD155 (poliovirus receptor). CD96 can reportedly mediate adhesive interactions between NK cells or T cells and CD155+ target cells. CD96 may compete with CD226 (DNAM 1) for CD155 binding and thus limit NK cell cytokine responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD96
Applications Flow Cytometry
Classification Monoclonal
Clone 6A6
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Cd96; T cell-activated increased late expression protein; Tactile; TACT
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Tactile Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.