Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cdkn2a, Rat, Clone: 5-C3-1, Abnova™

Catalog No. 89110998 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-998 Supplier Abnova Corporation Supplier No. MAB2417
Add to Cart
Add to Cart

Rat monoclonal antibody raised against synthetic peptide of Cdkn2a.

Specifications

Antigen Cdkn2a
Applications Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 5-C3-1
Conjugate Unconjugated
Description Rat monoclonal antibody raised against synthetic peptide of Cdkn2a.
Dilution Immunohistochemistry (Frozen sections) (5 ug/mL) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Cdkn2a
Gene Alias ARF-INK4a/Arf/INK4a-ARF/Ink4a/Arf/MTS1/Pctr1/p16/p16(INK4a)/p16INK4a/p19/p19ARF
Gene Symbols Cdkn2a
Host Species Rat
Immunogen A synthetic peptide corresponding to amino acids 54-75 of mouse Cdkn2a.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12578
Test Specificity This antibody is specific to residues 62-75 of murine p19ARF.
Target Species Mouse
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.