Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clasp2, Rat, Clone: KT68, Abnova™

Catalog No. 89113753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-753 Supplier Abnova Corporation Supplier No. MAB9738
Add to Cart
Add to Cart

Rat monoclonal antibody raised against partial recombinant Clasp2.

Specifications

Antigen Clasp2
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone KT68
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rat monoclonal antibody raised against partial recombinant Clasp2.
Dilution Western Blot (1 ug/mL) Immunofluorescence (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Clasp2
Gene Alias 1500004F14Rik/8030404L10Rik/C77448/CLASP2beta/mKIAA0627
Gene Symbols Clasp2
Host Species Rat
Immunogen Recombinant GST fusion protein corresponding to N-terminus of mouse Clasp2.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 76499
Target Species Mouse
Content And Storage Store at 4°C for 1 month. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.