Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF2, Rat, Biotin, Clone: BVD2-21C11, Abnova™

Catalog No. 89112495 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-495 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-495 Supplier Abnova Corporation Supplier No. MAB5863
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant CSF2.

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes and macrophages. The active form of the protein is found extracellularly as a homodimer. This gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q- syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. [provided by RefSeq

Specifications

Antigen CSF2
Applications ELISA
Classification Monoclonal
Clone BVD2-21C11
Conjugate Biotin
Description Rat monoclonal antibody raised against recombinant CSF2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CSF2
Gene Alias GMCSF/MGC131935/MGC138897
Gene Symbols CSF2
Host Species Rat
Immunogen Recombinant protein corresponding to human CSF2.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1437
Test Specificity human granulocyte/macrophage-colony stimulating factor (GM-CSF)1-4 The BVD2-21C11 antibody has been reported to cross react with GM-CSF from Rhesus monkey. (This is a neutralizing antibody.).
Target Species Human, Macaque
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.