Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSF3, Rat, Biotin, Clone: BVD11-37G10, Abnova™

Catalog No. 89112506 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-506 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-506 Supplier Abnova Corporation Supplier No. MAB5874
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant CSF3.

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes. The active protein is found extracellularly. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CSF3
Applications ELISA
Classification Monoclonal
Clone BVD11-37G10
Conjugate Biotin
Description Rat monoclonal antibody raised against recombinant CSF3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene CSF3
Gene Alias G-CSF/GCSF/MGC45931
Gene Symbols CSF3
Host Species Rat
Immunogen Recombinant protein corresponding to human CSF3.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1440
Test Specificity human Granulocyte-Colony Stimulating Factor (G-CSF)1,2. This is a neutralizing antibody.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.