Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ctcfl, Rat, Clone: KT72, Abnova™

Catalog No. 89108405 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-405 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-405 Supplier Abnova Corporation Supplier No. MAB10150
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Ctcfl.

Specifications

Antigen Ctcfl
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone KT72
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Ctcfl.
Dilution Western Blot (0.1-1 ug/mL) Immunofluorescence (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Ctcfl
Gene Alias Boris/OTTMUSG00000016680
Gene Symbols Ctcfl
Host Species Rat
Immunogen Mouse Ctcfl.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 664799
Target Species Mouse
Content And Storage Store at 4°C for one month. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.