Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-CYP11B1, Clone: 80-7,

Catalog No. MABS502MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS502MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS502MI Supplier MilliporeSigma™ Supplier No. MABS502
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects CYP11B1 clone: 80-7 in Human, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Cytochrome P450 11B1, mitochondrial (EC1.14.15.4, EC1.14.15.5; UniProt P15538; also known as CYPXIB1, Cytochrome P-450c11, Cytochrome P450C11, Cytochrome P450 subfamily XIB polypeptide 1, Cytochrome P450 XIB1, Steroid 11-beta-monooxygenase, Steroid 11-beta-hydroxylase) is encoded by the CYP11B1 (Also known as CPN1, CYP11B, FHI, P450C11, S11BH) gene in human (Gene ID 1584). CYP11B1 protein is a member of the cytochrome P450 superfamily of monooxygenases that catalyze reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. It is localized to the inner mitochondrial membrane and mediates the conversion of progesterone to cortisol in the adrenal cortex. CYP11B1 gene mutations are known causes of adrenal hyperplasia 4 (AH4), a form of congenital adrenal hyperplasia due to defective synthesis of cortisol.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP11B1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 80-7
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P15538
Gene Symbols CYP11B1; S11BH
Host Species Rat
Immunogen Chicken serum albumin-conjugated linear peptide corresponding to human CYP11B1 near the N-terminus.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000488
Target Species Human, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.