Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dendritic Cells Rat anti-Mouse, PE, Clone: 33D1, BD

Catalog No. BDB557578 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557578 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557578 Supplier BD Biosciences Supplier No. 557578
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 33D1 antibody reacts with an antigen on most dendritic cells (DC) of spleen, lymph node, and Peyer's patch, but not liver, bone marrow, or epidermal dendritic cells; macrophages; other leukocytes; or erythroid cells. Within the spleen, the majority of 33D1+ DC are localized in the marginal zones. Thymic dendritic cells may express a low level of the 33D1 antigen. It has been reported that bone-marrow DC can be induced to express the 33D1 antigen by culture in the presence of GM-CSF, and the resulting 33D1+ DC are effective in in vitro (induction of MLR) and in vivo (anti-tumoral vaccination) assays for antigen presentation. However, the addition of IL-4 to GM-CSF in bone-marrow cultures resulted in loss of 33D1 expression and enhanced the MLR-stimulatory activity of the DC. It has also been reported that 33D1 expression is upregulated when liver-derived DC are cultured on collagen-coated plates in the presence of GM-CSF. In vivo functional 33D1+ DC are induced in the brains of mice chronically infected with Toxoplasma gondii, probably via the parasite's induction of GM-CSF.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Dendritic Cells
Applications Flow Cytometry
Classification Monoclonal
Clone 33D1
Concentration 0.2mg/mL
Conjugate PE
Description Dendritic Cell inhibitory Receptor-2 (DCIR2)
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Dendritic cells purified from mouse spleen and lymph node
Purification Method Affinity Purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.