Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DYKDDDDK tag, Rat, Clone: L5, Abnova™

Catalog No. 89136340 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-340 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-340 Supplier Abnova Corporation Supplier No. MAB7373
Add to Cart
Add to Cart

Rat monoclonal antibody raised against DYKDDDDK tag.

Specifications

Antigen DYKDDDDK tag
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone L5
Conjugate Unconjugated
Description Rat monoclonal antibody raised against DYKDDDDK tag.
Dilution Immunofluorescence (1:10) Western Blot (1:1000) Immunocytochemistry (1:10) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Rat
Immunogen N-terminal DYKDDDDK-tagged extracellular domain of of mouse Langerin.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity DYKDDDDK Epitope Tag (L5).
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.