Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynll1, Rat, Clone: 10D6, Abnova™

Catalog No. 89111397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-397 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-397 Supplier Abnova Corporation Supplier No. MAB3403
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Dynll1.

Specifications

Antigen Dynll1
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 10D6
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Dynll1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Dynll1
Gene Alias Dlc8/Dnclc1/Pin
Gene Symbols Dynll1
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Dynll1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56455
Test Specificity Recognizes human and mouse DLC1. Does not cross-react with DLC2.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.