Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eng, Rat, Clone: MJ7/18, Abnova™

Catalog No. 89111298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-298 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-298 Supplier Abnova Corporation Supplier No. MAB3250
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Eng.

Specifications

Antigen Eng
Applications Flow Cytometry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone MJ7/18
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Eng.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Eng
Gene Alias AI528660/AI662476/CD105/S-endoglin
Gene Symbols Eng
Host Species Rat
Immunogen Native purified Eng from inflamed mouse skin.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13805
Test Specificity Mouse endoglin, Mr 90KDa.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.