Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rat anti-FcgRII/III Anibody, Clone: Ab93,

Catalog No. MABF838MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF838MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF838MI Supplier MilliporeSigma™ Supplier No. MABF838
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects FcgRII/III Anibody clone: Ab93 in Mouse samples, and it is validated for Activity Assay, Flow Cytometry, Immunocytochemistry

Murine low affinity IgG Fc region receptor FcγRII and FcγRIII (UniProt P08101 and P08508, respectively) are encoded by the Fcgr2 (also known as Fcgr2b, Ly-17) and Fcgr3 gene, respectively. Four different classes of IgG Fc receptors (FcγRs) have been reported: FcγRI/CD32b, FcγRII/CD16, FcγRIII/CD16.2, and FcγRIV/CD64. FcγRII, FcγRIII, and FcγRIV (UniProt P08101, P08508, and Q8R477, respectively) are low-affinity receptors for monomeric IgG, whereas FcγRI (UniProt P26151) is the only high-affinity FcγR. FcγRs play important roles in inflammatory cell activation, clearance, presentation of antigen, and maintenance of IgG homeostasis. In addition to binding immune complex (IC), FcγRs have been shown to bind to non-IgG ligands. For example, FcγRII is shown to bind oxLDL, while FcγRIII binding to an E. coli component negatively regulates the function of macrophage receptor with collagenous structure (MARCO). FcyRII activation is also reported to transduce a negative regulatory signal against CD38 cross-linking-induced proliferation of resting mature B cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcgRII/III Anibody
Applications Functional Assay, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone Ab93
Formulation Purified rat monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl without preservatives.
Gene Accession No. P08508; P08101
Gene Symbols Fcgr3; Fcgr2; Fcgr2b; Ly-17
Host Species Rat
Immunogen Whole cells expressing mouse FcgRII/III.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) CD32: NP_034317
Target Species Mouse
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.