Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glo1, Rat, Clone: 6F10, Abnova™

Catalog No. 89113032 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-032 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-032 Supplier Abnova Corporation Supplier No. MAB6768
Add to Cart
Add to Cart

Rat monoclonal antibody raised against full-length recombinant Glo1.

Specifications

Antigen Glo1
Applications ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6F10
Conjugate Unconjugated
Description Rat monoclonal antibody raised against full length recombinant Glo1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene Glo1
Gene Alias 0610009E22Rik/1110008E19Rik/2510049H23Rik/AW550643/GLY1/Glo-1/Glo-1r/Glo-1s/Glo1-r/Glo1-s/Qglo
Gene Symbols Glo1
Host Species Rat
Immunogen Recombinant GST fusion protein corresponding to full length mouse Glo1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109801
Test Specificity This antibody is specific to Glo1.
Target Species Human, Monkey, Mouse
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.