Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hsf1, Rat, Clone: 10H8, Abnova™

Catalog No. 89110904 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-904 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-904 Supplier Abnova Corporation Supplier No. MAB2285
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Hsf1.

Specifications

Antigen Hsf1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 10H8
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Hsf1.
Dilution Western Blot (1-2 ug/mL) Immunocytochemistry (10 ug/mL) Immunoprecipitation (12.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Hsf1
Gene Alias AA960185
Gene Symbols Hsf1
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Hsf1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15499
Test Specificity This antibody recognizes an ∼85 KDa protein (inactive form) in unstressed cell lysates and an ∼95 KDa protein (active form) in heat-shocked cell lysates.
Target Species Avian, Bovine, Guinea Pig, Hamster, Human, Monkey, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.