Learn More
CD294 (CRTH2) Rat anti-Human, Brilliant Violet 510, Clone: BM16, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740189
Description
The BM16 monoclonal antibody specifically binds to CD294. CD294 is encoded by PTGDR2 (Prostaglandin D2 receptor 2) and is also known as CRTH2 (chemoattractant receptor-homologous molecule expressed on Th2 cells), GPR44 (G protein-coupled receptor 44), DL1R, and DP2. CD294 is a member of the G protein-coupled leukocyte chemoattractant receptor family. CD294 is expressed on Th2 cells and type-2 innate lymphoid cells (ILC2), but not Th1 type cells. CD294 is detectable on CD4+ T cells in fresh PBMC but not on B cells and NK cells. CD294 is also expressed on peripheral blood basophils and eosinophils, suggesting its involvement allergic reactions. Phenotypic analysis of CD4+ T cells expressing CRTH2 demonstrated that they were also CD45RA-negative and CD45RO+ and CD25+. These cells produce Th2- but little or no Th1-type cytokines upon stimulation with PMA and Ionomycin.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

Specifications
CD294 (CRTH2) | |
Monoclonal | |
0.2mg/mL | |
CRTH2; PTGDR2 ; Prostaglandin D2 receptor 2; DL1R; DP2; GPR44 | |
Rat | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
BM16 | |
Brilliant Violet 510 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CRTH2 Transfected Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only