Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD294 (CRTH2) Rat anti-Human, Brilliant Violet 650, Clone: BM16, BD Optibuild™

Catalog No. BDB740616 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB740616 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB740616 Supplier BD Biosciences Supplier No. 740616
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The BM16 monoclonal antibody specifically binds to CD294. CD294 is encoded by PTGDR2 (Prostaglandin D2 receptor 2) and is also known as CRTH2 (chemoattractant receptor-homologous molecule expressed on Th2 cells), GPR44 (G protein-coupled receptor 44), DL1R, and DP2. CD294 is a member of the G protein-coupled leukocyte chemoattractant receptor family. CD294 is expressed on Th2 cells and type-2 innate lymphoid cells (ILC2), but not Th1 type cells. CD294 is detectable on CD4+ T cells in fresh PBMC but not on B cells and NK cells. CD294 is also expressed on peripheral blood basophils and eosinophils, suggesting its involvement allergic reactions. Phenotypic analysis of CD4+ T cells expressing CRTH2 demonstrated that they were also CD45RA-negative and CD45RO+ and CD25+. These cells produce Th2- but little or no Th1-type cytokines upon stimulation with PMA and Ionomycin.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD294 (CRTH2)
Applications Flow Cytometry
Classification Monoclonal
Clone BM16
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Description CRTH2; PTGDR2 ; Prostaglandin D2 receptor 2; DL1R; DP2; GPR44
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Human CRTH2 Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.