Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stenotrophomonas maltophilia, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89138008 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-008 Supplier Abnova Corporation Supplier No. PAB14747

Rat polyclonal antibody raised against Stenotrophomonas maltophilia.

Specifications

Antigen Stenotrophomonas maltophilia
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rat polyclonal antibody raised against Stenotrophomonas maltophilia.
Dilution ELISA (1:2000-1:10000) Western Blot (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Host Species Rat
Immunogen Stenotrophomonas maltophilia
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 50μL of distilled water and 50μL of glycerol, store at -20°C for 2 years. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.