Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Icam1, Rat, Clone: BE29G1, Abnova™

Catalog No. 89111697 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-697 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-697 Supplier Abnova Corporation Supplier No. MAB4107

Rat monoclonal antibody raised against Icam1.

Specifications

Antigen Icam1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone BE29G1
Conjugate Unconjugated
Description intercellular adhesion molecule 1
Formulation In 10mM PBS, pH 7.2 (1% BSA, 0.09% sodium azide)
Gene Icam1
Gene Alias CD54, Icam-1, Ly-47, MALA-2, MGC6195
Gene Symbols Icam1
Host Species Rat
Immunogen Mouse Icam1.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15894
Test Specificity Anti-Mouse CD54 (ICAM-1) recognizes the 95KDa cell surface glycoprotein known as intracellular adhesion molecule ICAM-1. ICAM-1 is the receptor for LFA-1 and Mac-1.1, 2, 3. Anti-CD54 blocks the binding of LFA-1 to ICAM-1. The CD54 is present on endothelial cells, lymphocytes, epithelial cells, dendritic cells and keratinocytes. It has been demonstrated that ICAM-1 enhances T-cell activation.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.