Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Icam2, Rat, Clone: 3C4, Abnova™

Catalog No. 89111306 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-306 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-306 Supplier Abnova Corporation Supplier No. MAB3267
Add to Cart
Add to Cart

Rat monoclonal antibody raised against Icam2.

Specifications

Antigen Icam2
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone 3C4
Conjugate Unconjugated
Description Rat monoclonal antibody raised against Icam2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Icam2
Gene Alias CD102/Icam-2/Ly-60
Gene Symbols Icam2
Host Species Rat
Immunogen COS cells transfected with mouse ICAM-2 cDNA.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15896
Test Specificity Mouse ICAM-2.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.