Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ifng, Rat, Biotin, Clone: XMG1.2, Abnova™

Catalog No. 89112519 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-519 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-519 Supplier Abnova Corporation Supplier No. MAB5887
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Ifng.

Specifications

Antigen Ifng
Applications ELISA
Classification Monoclonal
Clone XMG1.2
Conjugate Biotin
Description Rat monoclonal antibody raised against recombinant Ifng.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Ifng
Gene Alias IFN-g/IFN-gamma/Ifg
Gene Symbols Ifng
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Ifng.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15978
Test Specificity Mouse Interferon-gamma (IFN-gamma)1-3. This is a neutralizing antibody.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.