Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Fc, Rat, Clone: KT46, Abnova™

Catalog No. 89136760 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-760 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-760 Supplier Abnova Corporation Supplier No. MAB9724
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant human IgG1 Fc.

Specifications

Antigen IgG1 Fc
Applications ELISA
Classification Monoclonal
Clone KT46
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant human IgG1 Fc.
Dilution ELISA (1 ug/mL) Sandwich ELISA (5 ug/mL for coating) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Host Species Rat
Immunogen Recombinant fusion protein corresponding to human IgG1 Fc.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific to human IgG1. Not cross-react with human IgG2, IgG3 and IgG4, IgA, IgE and IgM.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.