Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL10, Rat, Clone: JES3-9D7, Abnova™

Catalog No. 89110697 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-697 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 89-110-697 Supplier Abnova Corporation Supplier No. MAB1896
This item is not returnable. View return policy

Rat monoclonal antibody raised against recombinant IL10.

The protein encoded by this gene is a cytokine produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. This cytokine can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice suggested the function of this cytokine as an essential immunoregulator in the intestinal tract. [provided by RefSeq

Specifications

Antigen IL10
Applications ELISA, Flow Cytometry, Inhibition Assays
Classification Monoclonal
Clone JES3-9D7
Conjugate Unconjugated
Description interleukin 10
Formulation In PBS, pH 7.2
Gene IL10
Gene Alias CSIF, IL-10, IL10A, MGC126450, MGC126451, TGIF
Gene Symbols IL10
Host Species Rat
Immunogen Recombinant protein corresponding to human IL10.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3586
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.