Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL10, Rat, Clone: MI10-L2, Abnova™

Catalog No. 89110359 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-359 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-359 Supplier Abnova Corporation Supplier No. MAB1322
Add to Cart
Add to Cart

Rat monoclonal antibody raised against full-length recombinant Il10.

Specifications

Antigen IL10
Applications ELISA, Western Blot
Classification Monoclonal
Clone MI10-L2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against full length recombinant Il10.
Dilution ELISA (1:2000) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Il10
Gene Alias CSIF/Il-10
Gene Symbols Il10
Host Species Rat
Immunogen Recombinant protein corresponding to full length mouse Il10.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16153
Test Specificity React with mouse Il10. Does not crossreact with human IL10.
Content And Storage Store at 4°C on dry atmosphere. After reconstitutingstore at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.