Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Il12b, Rat, Polyclonal Antibody, Abnova™

Catalog No. 89119617 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-119-617 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-119-617 Supplier Abnova Corporation Supplier No. PAB18026

Rat polyclonal antibody raised against recombinant Il12b.

Specifications

Antigen Il12b
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rat polyclonal antibody raised against recombinant Il12b.
Dilution ELISA (1:5000) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS.
Gene Il12b
Gene Alias Il-12b/Il-12p40/Il12p40/p40
Gene Symbols Il12b
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Il12b.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16160
Test Specificity In ELISA, this antibody shows reactivity with IL-12 p40 and no cross-reactivity with murine IL-10 and other tested proteins.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.