Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL27, Rat, Clone: 3H12.F10, Abnova™

Catalog No. 89108467 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-467 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-467 Supplier Abnova Corporation Supplier No. MAB10274
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Il27.

Specifications

Antigen IL27
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 3H12.F10
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Il27.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Il27
Gene Alias IL-27/IL-27p28/Il30/p28
Gene Symbols Il27
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Il27.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246779
Test Specificity This antibody is specific for mouse and rat p28 protein. Cross-reactivity with IL-27 from other sources has not been determined.
Target Species Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.