Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL2ra/b/g, Rat, Clone: IR2-D4, Abnova™

Catalog No. 89110371 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-371 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-371 Supplier Abnova Corporation Supplier No. MAB1351
Add to Cart
Add to Cart

Rat monoclonal antibody raised against native Il2ra/b/g.

Specifications

Antigen IL2ra/b/g
Applications Flow Cytometry
Classification Monoclonal
Clone IR2-D4
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native Il2ra/b/g.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Alias CD25/Il2r/Ly-43/MGC130561
Gene Symbols Il2ra
Host Species Rat
Immunogen Native from Con-A activated T cells.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16184/16185/16186
Test Specificity React with mouse Il2 Receptor. Chain specificity was not determined.
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.