Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL3, Rat, Clone: BVD8-3G11, Abnova™

Catalog No. 89111318 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-318 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-318 Supplier Abnova Corporation Supplier No. MAB3291
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant IL3.

The protein encoded by this gene is a potent growth promoting cytokine. This cytokine is capable of supporting the proliferation of a broad range of hematopoietic cell types. It is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. This cytokine has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders. [provided by RefSeq

Specifications

Antigen IL3
Applications Western Blot
Classification Monoclonal
Clone BVD8-3G11
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant IL3.
Dilution Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene IL3
Gene Alias IL-3/MCGF/MGC79398/MGC79399/MULTI-CSF
Gene Symbols IL3
Host Species Rat
Immunogen Recombinant protein corresponding to human IL3.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3562
Test Specificity human Interleukin-3 (IL-3). This is a neutralizing antibody.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.