Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL3, Rat, Clone: MP2-8F8, Abnova™

Catalog No. 89111339 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-339 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-339 Supplier Abnova Corporation Supplier No. MAB3314
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant Il3.

Specifications

Antigen IL3
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone MP2-8F8
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant Il3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Il3
Gene Alias BPA/Csfmu/HCGF/Il-3/MCGF/PSF
Gene Symbols Il3
Host Species Rat
Immunogen Recombinant protein corresponding to mouse Il3.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16187
Test Specificity Mouse Interleukin-3.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.