Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL4, Rat, Clone: I4-F5, Abnova™

Catalog No. 89110362 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-362 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-362 Supplier Abnova Corporation Supplier No. MAB1325
Add to Cart
Add to Cart

Rat monoclonal antibody raised against full-length recombinant Il4.

Specifications

Antigen IL4
Applications ELISA
Classification Monoclonal
Clone I4-F5
Conjugate Unconjugated
Description Rat monoclonal antibody raised against full length recombinant Il4.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene Il4
Gene Alias Il-4
Gene Symbols Il4
Host Species Rat
Immunogen Recombinant protein corresponding to full length mouse Il4.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16189
Test Specificity React with mouse Il4. Does not cross react with rat or human IL4.
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.