Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL4, Rat, Clone: MP4-25D2, Abnova™

Catalog No. 89111320 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-320 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-320 Supplier Abnova Corporation Supplier No. MAB3293
Add to Cart
Add to Cart

Rat monoclonal antibody raised against recombinant IL4.

The protein encoded by this gene is a pleiotropic cytokine produced by activated T cells. This cytokine is a ligand for interleukin 4 receptor. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play a central role in mediating the immune regulatory signal of this cytokine. This gene, IL3, IL5, IL13, and CSF2 form a cytokine gene cluster on chromosome 5q, with this gene particularly close to IL13. This gene, IL13 and IL5 are found to be regulated coordinately by several long-range regulatory elements in an over 120 kilobase range on the chromosome. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq

Specifications

Antigen IL4
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone MP4-25D2
Conjugate Unconjugated
Description Rat monoclonal antibody raised against recombinant IL4.
Dilution Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene IL4
Gene Alias BCGF-1/BCGF1/BSF1/IL-4/MGC79402
Gene Symbols IL4
Host Species Rat
Immunogen Recombinant protein corresponding to human IL4.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3565
Test Specificity human Interleukin-4 (IL-4) The MP4-25D2 antibody has been reported to cross react with IL-4 from Rhesus monkeys. This antibody has also been reported to be useful for epitope mapping of human IL-4.2 This is a neutralizing antibody.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.